Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Hydroxide (Powder/USP/FCC), Fisher Chemical

Catalog No. M342500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M342-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M342-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
IUPAC Name magnesium(2+) dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 350°C
Color White
pH 9.5 to 10.5
Physical Form Solid
Quantity 500 g
Linear Formula Mg(OH)2
Formula Weight 58.32
Percent Purity 95.0 to 100.5% (FCC)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP)
Loss on Drying 2.0% max. (FCC), 2.0% max. (FCC/USP), 2.0% max. (USP)
Loss on Ignition 30 to 33% (FCC), 30 to 33% (FCC/USP), 30 to 33% (USP)
Packaging Poly Bottle
Chemical Name or Material Magnesium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.