Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Magnesium Metal (Ribbon/Turnings)
SCI_ED

Catalog No. S25397 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2.5 g
50 g
Packaging:
Lab Grade - Ribbon
Turnings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25397 50 g Lab Grade - Ribbon
S25397A 2.5 g Turnings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25397
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silver-white magnesium ribbon for classroom demonstrations and lab experiments. Ideal for illustrating reactivity and combustion reactions.

  • Specifically packaged for instructional use in chemistry labs
  • Ideal for demonstrating chemical reactivity, including combustion and single displacement reactions
  • Ribbon/turnings format for easy handling and clean ignition
  • Lab-grade material for consistent performance
  • 50g size is suitable for repeated classroom use
  • Color: Silver-white
  • Density: 1.74g/mL
  • Formula: Mg
  • Formula Weight: 24.31
  • Lab-grade packaging ensures safe and convenient storage
  • For educational use only; Certificate of Analysis (C of A) not available

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Color Silver-White
Density 1.74g/mL
Quantity 50 g
Packaging Lab Grade - Ribbon
Linear Formula Mg
Formula Weight 24.31
Grade Reagent
Chemical Name or Material Magnesium Metal
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 4.1, III, : Flammable Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.