Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium molybdenum oxide, 99.9% (metals basis)

Catalog No. AA3937618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3937618 50 g
AA3937630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3937618 Supplier Thermo Scientific Chemicals Supplier No. 03937618
Add to Cart

CAS: 13767-03-8 | MgMoO4 | 184.251 g/mol

Magnesium molybdenum oxide is used as an intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13767-03-8
Molecular Formula MgMoO4
Molecular Weight (g/mol) 184.251
MDL Number MFCD00016195
InChI Key MODMKKOKHKJFHJ-UHFFFAOYSA-N
Synonym magnesium molybdate, magnesium molybdenum oxide, unii-ou5m6mzi9r, ou5m6mzi9r, magnesium dioxido dioxo molybdenum, mg.moo4, magnesium-molybdate, magnesium 2+ molybdate, magnesium 2+ ion molybdate
PubChem CID 3084170
IUPAC Name magnesium;dioxido(dioxo)molybdenum
SMILES [O-][Mo](=O)(=O)[O-].[Mg+2]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 184.25
Percent Purity 99.9%
Odor Odorless
Density 2.208 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium molybdenum oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 237-376-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.