Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium oxide, 97+%, for analysis

Catalog No. AC222535000
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222535000 500 g Glass bottle
AC222532500 250 g Glass bottle
AC222530010 1 kg Plastic bottle
AC222531000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC222535000 Supplier Thermo Scientific Chemicals Supplier No. 222535000

CAS: 1309-48-4 | MgO | 40.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Color White
Physical Form Amorphous Powder
Quantity 500 g
Assay Percent Range 97+%
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol
Formula Weight 40.29
Percent Purity ≥97.0%
Grade Analytical
Loss on Ignition 3.0% max.
Packaging Glass bottle
Density 3.5800g/mL
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.