Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxide, Heavy Powder, USP, 96-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604078 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-078 500 g Poly Bottle
18-604-079 2.5 kg Poly Pail
18-604-081 25 kg Fiber Drum
18-604-080 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-078 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA125500GM
Add to Cart
Add to Cart

MA125, 1309-48-4, MgO

Spectrum™ Chemical Magnesium Oxide, Heavy Powder, USP has many uses, but from a pharmaceutical perspective it is used primarily as a mineral supplement and a tablet and capsule diluent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 96 to 100.5%
Grade USP
Loss on Ignition 10%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.