Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Oxide, Powder, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18604089 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-089 500 g Poly Bottle
18-604-094 5 g Amber Glass Bottle
18-604-086 25 g Poly Bottle
18-604-087 125 g Poly Bottle
18-604-088 6 x 125 g Poly Bottle
18-604-091 2.5 kg Poly Pail
18-604-090 6 x 500 g Poly Bottle
18-604-092 12 kg Poly Pail
18-604-093 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-604-089 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1055500GM
Add to Cart
Add to Cart

M1055, 1309-48-4, MgO

Spectrum™ Chemical Magnesium Oxide, Powder, Reagent, ACS is a white solid mineral that is found naturally as periclase and is considered a source of magnesium. It is also considered a metastable compound. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 95%
Grade ACS
Loss on Ignition 2%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.