Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Phosphate, Dibasic, Trihydrate, FCC, 96%, Spectrum™ Chemical
SDP

Catalog No. 18604095 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-095 500 g Poly Bottle
18-604-096 2.5 kg Poly Pail
18-604-097 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-095 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA127500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

MA127, 7782-75-4, MgHPO4·3H2O

Spectrum™ Chemical Magnesium Phosphate, Dibasic, Trihydrate, FCC is listed on the FDA′s Generally recognized as safe, or GRAS, list of substances. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
IUPAC Name magnesium(2+) trihydrate hydrogen phosphate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 96%
Grade FCC
Loss on Ignition 29.0-36%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.