Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium powder, -100+200 mesh, 99.6% (metals basis excluding Ca), Thermo Scientific Chemicals

Catalog No. AA3619430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3619430 250 g
AA3619418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3619430 Supplier Thermo Scientific Chemicals Supplier No. 03619430
Add to Cart

missing translation for 'validMainframeMsg'

For vacuum deposition

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Color White
Quantity 250 g
Mesh Size −100+200 Mesh
Odor Odorless
Form Powder
UN Number UN1418
Percent Purity 99.6%
Assay (metals basis excluding Ca), Ca typically 0.1-1%
Chemical Name or Material Magnesium powder
Hazard Category H250-H260-H500
Hazard Statement GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: MAGNESIUM, POWDER
EINECSNumber 231-104-6
RTECSNumber OM2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.