Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Magnesium Reagents for Hitachi™ Analyzers

Catalog No. 23666366 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
4 x 50mL R1, 4 x 50mL R2
5 x 250 R1mL
5 x 250 R2mL
Formulation:
DMSO (Dimethyl Sulfoxide) 20%, Xylidyl blue
R1: KCN 0.02% w/v, EGTA
Xylidyl blue 0.1mM, EGTA 0.13mM, DMSO 1.4M, Buffer, Surfactant, non-reactive Stabilizers including Potassium Cyanide at 0.02% w/v
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
23-666-366 5 x 250 R1mL R1: KCN 0.02% w/v, EGTA
23-666-365 4 x 50mL R1, 4 x 50mL R2 Xylidyl blue 0.1mM, EGTA 0.13mM, DMSO 1.4M, Buffer, Surfactant, non-reactive Stabilizers including Potassium Cyanide at 0.02% w/v
23-666-367 5 x 250 R2mL DMSO (Dimethyl Sulfoxide) 20%, Xylidyl blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-366 Supplier Pointe Scientific Supplier No. HM429R1

Includes

Reagent 1 and Reagent 2 when combined: Xylidyl blue 0.1mM; EGTA 0.13mM; DMSO 1.4M; Buffer; surfactant; nonreactive stabilizers including potassium cyanide at 0.02% w/v

Requires

Calibrator

Quantify magnesium levels in serum

  • Method: Xylidyl Blue, Endpoint
  • Format: Liquid (ready to use)
  • Linearity: 4.86mg/dL
  • Expected Values: 1.6 to 3.0mg/dL

Applications:

For the quantitative determination of magnesium in serum with Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Magnesium R1
Formulation R1: KCN 0.02% w/v, EGTA
For Use With (Application) Quantitative determination of Magnesium in Serum
Quantity 5 x 250 R1mL
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.