Learn More
Magnesium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Shop All Fisherbrand ProductsSpecifications
Specifications
| Packaging | Poly Bottle |
| Vapor Pressure | 14mmHg at 20°C |
| Physical Form | Liquid |
| Concentration | 990 to 1010 ppm |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Quantity | 100 mL |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 1000ppm ±1% |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 8, : Corrosive |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.