Learn More
Magnesium silicide, 99.99% (metals basis)
Description
Magnesium silicide is used to produce alloys such as aluminum alloys. It is also used in food, beverages, as a flavor enhancer and as an intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1102°C |
| Color | Blue |
| Physical Form | Pieces |
| Quantity | 50 g |
| Assay Percent Range | 99.99% (Metals basis) |
| Solubility Information | Insoluble in water and denser than water. |
| Formula Weight | 76.71 |
| Sensitivity | Air and moisture sensitive |
| Density | 2 |
| Chemical Name or Material | Magnesium silicide |
Safety and Handling
| Hazard Category | H261 |
| Precautionary Statement | P223-P231+P232-P280-P335+P334-P370+P378q-P501c |
| DOTInformation | DOT Class: 4.3, Packaging Group: II |
| EINECSNumber | 245-254-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.