Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate, Anhydrous, Powder, CP, Spectrum™ Chemical
SDP

Catalog No. 18604122 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
110 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-122 2.5 kg Poly Pail
18-604-121 500 g Poly Bottle
18-604-124 110 lb. Fiber Drum
18-604-123 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-122 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M10642.5KG
Add to Cart
Add to Cart

M1064, 7487-88-9, MgSO4

Spectrum™ Chemical Magnesium Sulfate, Anhydrous, Powder, CP is hygroscopic and used as a drying agent. CP or Chemically Pure grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Grade Chemically Pure
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.