Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Solstice

Catalog No. 6001967 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
5 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-67 100 g
60-019-68 1 kg
60-019-70 5 kg
60-044-774 6 x 1 kg
60-044-775 6 x 2.5 kg
60-044-773 4 x 5 kg
60-019-69 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60-019-67 Supplier Solstice Supplier No. 13142100G

Meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 100 g
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
Formula Weight 246.47g/mol
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Packaging Plastic Bottle
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
EINECSNumber 231-298-2
RTECSNumber OM4508000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.