Magnesium trisilicate, Alfa Aesar™

Manufacturer: Alfa Aesar 33338A1

Quantity 1kg

 View more versions of this product

Catalog No. AA33338A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14987-04-3
    Physical Form Powder
    Molecular Formula Mg2O8Si3
    Formula Weight 260.86
    MDL Number MFCD00016203
    Synonym Magnesium silicate
    Chemical Name or Material Magnesium trisilicate

    Specifications

    Physical Form Powder
    Formula Weight 260.86
    Color White
    Quantity 1kg
    EINECS Number 239-076-7
    Molecular Formula Mg2O8Si3
    MDL Number MFCD00016203
    CAS 14987-04-3
    Synonym Magnesium silicate
    Chemical Name or Material Magnesium trisilicate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit