Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Magnetic Antibody Labeling Kit
SDP

Catalog No. 13000015
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-000-015 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-000-015 Supplier Novus Biologicals™ Supplier No. 13000015

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Antibody labeling kit

The Magnetic Conjugation Kit allows one-step conjugation of antibodies, proteins and peptides (or any other biomolecule with an amine group) to specially treated magnetic particles without the need for extensive optimization of the conjugation reaction.

Kit Contents:

  • 1 Midi vial of 500 nm (0.5 μm) Magnetic particles
  • 1 bottle of Mag Storage Buffer
  • 1 vial/bottle of Mag Wash Buffer
  • 1 vial/bottle of Mag Antibody Diluent
  • 1 vial/bottle of Mag Quencher

Specifications

Quantity 1 mg
Product Type Antibody Labeling Kit
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.