Learn More
V&P Scientific Inc MAGNETIC BEAD SEPARATION BASE, Base for the VP 772F2R Rack Series
Supplier:  V&P Scientific Inc VP 772F2M1
Encompass
MAGNETIC BEAD SEPARATION BASE, Base for the VP 772F2R Rack Series, 1 Sided Bead Pellet Location, High Position, 48MGO, NdFeB Magnet, Polycarbonate
Catalog No. NC0542874
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.