Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Magnetic Field Sensor
SDPSCI_ED

Catalog No. S16064ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
K-8, High School, College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16064ND K-8, High School, College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16064ND Supplier Vernier™ Supplier No. MGBTA
Add to Cart
Add to Cart

Includes

Vernier Magnetic Field Sensor

Requires

Requires an interface and software, sold separately

The Magnetic Field Sensor can be used to study the field around permanent magnets, coils, and electrical devices. It features a rotating sensor tip to measure both transverse and longitudinal magnetic fields.

Typical Resolution ± 0.32 mT range: 0.0002 mT ± 6.4 mT range: 0.004 mT

Warranty and Services

5-year limited warranty

Specifications

Includes Rotating sensor tip
Grade K-8, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution ±0.32 mT range: 0.0002 mT; ±6.4 mT range: 0.004 mT
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.