Learn More
Brady™ Magnetic Non-Lockable Confined Space Training Kits
Catalog No. 19129764 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
Confined space. Enter by permit only.
Permit required. No unauthorized entry.
Dimensions (L x W):
20 in.
28 in.
34 in.
40 in.
50 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Dimensions (L x W)
19-129-764 Permit required. No unauthorized entry. 34 in.
19-129-759 Confined space. Enter by permit only. 20 in.
19-129-760 Permit required. No unauthorized entry. 20 in.
19-129-761 Confined space. Enter by permit only. 28 in.
19-129-762 Permit required. No unauthorized entry. 28 in.
19-129-763 Confined space. Enter by permit only. 34 in.
19-129-765 Confined space. Enter by permit only. 40 in.
19-129-766 Permit required. No unauthorized entry. 40 in.
19-129-767 Confined space. Enter by permit only. 50 in.
19-129-768 Permit required. No unauthorized entry. 50 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-129-764 Supplier Brady™ Supplier No. 151074
Visually identifying your confined spaces will keep you compliant, but it may leave gaps in safety. Add another level of safety with this Confined Space Safety Cover to keep your crew safe.
  • Get the complete safety package – unlike signs and tapes this cover provides a sign and an unavoidable boundary in one easy-to-use product
  • The most versatile option available – magnetic covers easily manipulated to fit a variety of opening sizes and shapes, including hinged manways and door sheets
  • Make it last – unlike wasteful warning tape, these covers are designed to be reused for multiple projects
Specifications
Product Type Confined Space Cover
Color Red
Material Polyester
Dimensions (L x W) 34 in.
Displayed Text Permit required. No unauthorized entry.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.