Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Magnetic Stir Bars With Pivot Ring
SDPSCI_ED

Catalog No. S111104 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Diameter (Metric):
4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S111104 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111104 Supplier United Scientific™ Supplier No. MSP155

These magnetic stir bars are constructed of high quality magnets coated with PTFE.

  • Octagon-shaped magnetic stir bar with pivot ring
  • High-quality magnet coated with PTFE

Specifications

For Use With (Application) Stirring
Material PTFE Coated Magnets
Dimensions (L x W x H) 15 x 4 x 4 mm
Autoclavable Autoclavable
Coating PTFE
Diameter (Metric) 4 mm
Shape Octagon With Pivot Ring
Type Stir Bar
Quantity 1 Pk.
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.