Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Magnetic Stir Bars Without Pivot Ring
SDPSCI_ED

Catalog No. S111105 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
3 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S111105 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111105 Supplier United Scientific™ Supplier No. MSZ20
Add to Cart
Add to Cart

These magnetic stir bars are constructed of high quality magnets coated with PTFE.

  • Polygon-shaped magnetic stir bar without pivot ring
  • High-quality magnet coated with PTFE

Specifications

Length (Metric) 3 mm
Material PTFE, Magnetic Alloy
For Use With (Application) Stirring
Coating PTFE
Diameter (Metric) 3 mm
Quantity 1 Ea.
Shape Polygon
Autoclavable Autoclavable
Color White
Dimensions (L x W x H) 20 x 3 x 3 mm
Type Stir Bar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.