Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Magnetic Wand Retriever
SCI_ED

Catalog No. S89505 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89505 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89505 Supplier Eisco™ Supplier No. MAGWAND
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Magnetic Wand Retriever picks up light magnetized objects easily and safely.

  • Perfect for easy cleanup of magnetized objects such as stir bars, iron filings, paper clips and screws.
  • Colorful and safe hands-on tool for exploring the properties of magnets and magnetism.
  • Plastic wand encases a magnet in a 2.3 x 1 x 0.5 in. housing on one end.
  • Easy grip 5 in. long handle has a loop hole for hanging storage.

Specifications

Material Plastic
Shape Rectangular
Grade 9 to 12
Color Purple
Product Type Magnet
Dimensions (L x W x H) 7.3 x 1 x 0.5 in.
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.