Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Magnetic Wands
SDPSCI_ED

Catalog No. S00664 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00664 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00664 Supplier United Scientific™ Supplier No. UNMGTWNDPK/6
Add to Cart
Add to Cart
This item is not returnable. View return policy

Magentic Wands are a classic and simple way to demonstrate magnetism.

  • These wands have many uses for your classroom.
  • Use it to show what items are attracted to a magnet, which proves it contain cobalt, nickel, or iron.
  • Comes as a pack of 6 in various colors.

Specifications

Grade 6 to 12
Format Set
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.