Learn More
Description
- For performing manual wash procedure with assays containing MagPlex™ Microparticles
- The side clasps on this magnet have been specially designed to hold a round-bottom plate securely to the magnet, enabling the user to quickly decant it over a waste container
- Similar to the method used in an ELISA, this manual plate washing method has been shown to effectively remove nearly all of the supernatant from every well on the plate, with virtually no bead carry-over
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Instrument |
| For Use With (Application) | Performing manual wash procedure with assays containing MagPlex Microparticles |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
