Learn More
Cytiva MagRack Maxi
Catalog No. 45002805 Shop All Cytiva Products
Change view
Click to view available options
Material:
Anodized Aluminum, Plastic
Catalog No. Material
45-002-805 Anodized Aluminum, Plastic
1 options
Catalog No. 45-002-805 Supplier Cytiva Supplier No. 28986441
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
A magnetic rack for small-scale protein purification and sample enrichment with magnetic beads
For use with Protein A Mag Sepharose, Protein A Mag Sepharose Xtra, Protein G Mag Sepharose, Protein G Mag Sepharose Xtra, His Mag Sepharose Ni, NHS Mag Sepharose, TiO2 Mag Sepharose, and Streptavidin Mag Sepharose.
  • Anodized aluminum housing (blue) with a detachable plastic bar (white) containing a neodymium magnet
  • Covers a broad range of sample volumes from low–microliter to high–milliliter purification scales
  • Ability to vary the amount of medium as well as the sample volume
  • Enrichment of sample volumes up to 50mL (1.7 oz.)
  • Performs small-scale purifications
TRUSTED_SUSTAINABILITY
Specifications
Material Anodized Aluminum, Plastic