Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAK10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565672 Supplier Invitrogen™ Supplier No. PA565672
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDDDSGWELSM PEKMEKSNTN WVDITQDFEE ACRELKLGEL LHDKLFGLFE AMSAIEMMDP KMDAGMIGNQ VNRKVL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

The MAK10 gene encodes a 733-amino acid protein with several regions of similarity to T cell receptor alpha-subunit V (variable) regions in yeast. The mammalian homologue of yeast MAK10, also known as EGAP, is one subunit of a novel N-terminal acetyltransferase (NAT) that is highly conserved among vertebrate species. It is expressed in a variety of tissues in the developing rat embryo but restricted in expression in the adult, remaining detectable only in tissues undergoing continual cell renewal or in cells responding to pathological injury. The MAK10-NAT complex is an essential regulatory enzyme controlling the function of a subset of proteins required for embryonic growth control and vessel development. This complex functionally co-assembles in mammalian cells to regulate cell proliferation and is essential for embryonic development, at least in part through the regulation of target of rapamycin (TOR) signaling events. At least two isoforms of MAK10 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAK10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAA35
Gene Accession No. Q5VZE5
Gene Alias A330021G12Rik; A330027C19Rik; AI158944; bA379P1.1; C030004C14Rik; corneal wound healing related protein; corneal wound healing-related protein; corneal wound-healing-related protein; EGAP; Emb8; Embryonic growth-associated protein; embryonic growth-associated protein homolog; Mak10; MAK10 homolog, amino-acid N-acetyltransferase subunit; MAK10P; N(alpha)-acetyltransferase 35, NatC auxiliary subunit; NAA35; Nalpha acetyltransferase 35; N-alpha-acetyltransferase 35, NatC auxiliary subunit; protein MAK10 homolog; RAT52; RP11-379P1.1; T4a
Gene Symbols NAA35
Host Species Rabbit
Immunogen Recombinant Human MAK10. Recombinant protein control fragment (Product #RP-104926).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60560
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.