Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAK16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120695 Supplier Invitrogen™ Supplier No. PA5120695
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver, Mouse testis, Rat testis. Immunogen sequence: RNEYSLTGLC NRSSCPLANS QYATIKEEKG QCYLYMKVIE RAAFPRRLWE RVRLSKNYEK ALEQIDENLI YWPRFIRHKC KQRFTKITQY L.

MAK16 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAK16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mak16
Gene Accession No. Q8BGS0, Q9BXY0
Gene Alias 2600016B03Rik; AI314911; MAK16; MAK16 homolog; MAK16 homolog (S. cerevisiae); MAK16L; NNP78; Protein MAK16 homolog; Protein RBM13; Rbm13; RGD1311297; RNA binding motif protein 13; RNA binding protein
Gene Symbols Mak16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-120 of human MAK16 (NP_115898.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306526, 67920, 84549
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.