Learn More
Medchemexpress LLC HY-103663 5mg Medchemexpress, MAK683 CAS:1951408-58-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1036635MG
Encompass_Preferred
Medchemexpress, HY-103663 5mg MAK683 CAS:1951408-58-4 MAK683 is an embryonic ectoderm development (EED) inhibitor extracted from patent US20160176882 A1, compound example 2; has IC50s of 59, 89, 26 nM in EED Alphascreen binding, LC-MS and ELISA assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8154
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.