Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Making a Wire Lithograph & Pressure Sensor Nanotechnology STEM Kit
SDPSCI_ED

Catalog No. S43773 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
9 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43773 9 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43773 Supplier United Scientific Supplies Supplier No. AISNT1

missing translation for 'validMainframeMsg'

Developed with Argonne National Laboratory, this kit gives students the opportunity to apply a revolutionary nanotechnology manufacturing technique to grow nano-scale and micro-scale copper wires.

  • Developed with Argonne National Laboratory, this kit gives students the opportunity to apply a revolutionary nanotechnology manufacturing technique to grow nano-scale and micro-scale copper wires.
  • Using a unique ultra nanocrystalline diamond (UNCD) electrode, students use traditional electroplating and lithography (E&L) techniques to grow pattern-shaped wires that become the essential components that can be used to fabricate optical and electrical sensors and other devices.
  • This kit covers the following concepts:
  • Measurement scales / perspectives
  • Unique properties at nanoscale
  • Fabricating nano / microscale structures
  • Creating nano / microscale lithographic wire patterns
  • Designed for use by 40 students working in groups of four.
  • Kit is provided with an access code for downloadable activity guides and other support information, including:
  • Teacher Guide
  • Student Guide
  • Glossary
  • Background Information: Learning About Nanotechnology
  • Videos: Nano Frontiers MP4, Nano Manufacturing: Electroplate and Lift Lithography – Making Nanowires mp4
  • Powerpoints: Making a Microwire Lithograph pp/mp4, Nano Technology: A Closer Look at the Universe of little ppt/mp4
  • Poster: A Closer Look at Nanowire

Specifications

Grade 9 to 15
Format Kit
Product Type Nanotechnology Kit
Languages English
Subject STEM, Physics - Nanotechnology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.