Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BlueMed MAKRO Reusable Shoe Covers

Catalog No. 18009162
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-009-162 Large
18-009-160 Small
18-009-161 Medium
18-009-163 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-009-162 Supplier BlueMed Supplier No. RES1803P
This item is not returnable. View return policy

BlueMed MAKRO reusable shoe covers are washable, anti-skid, waterproof, low linting and static dissipating

  • Super Anti-Skid Sole
  • Waterproof
  • Static Dissipating
  • High Quality with Double Stitched and Reinforced Seams
  • Ankle High
  • Durable - 6 to 9 month lifetime
  • Reusable - Washable with Cold Water, Air Dry
  • Made in the USA

Specifications

Size Large
Material Polyester
Latex Free Yes
Color Gunmetal
Disposable Reusable
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-skid
Model RES-1803-P
Seam Type Double-stitched and reinforced seams
Packaging Quantity 1 Pair
Packaging Type Bags
Length (English) 14 in.
Length (Metric) 35.5 cm
Height (English) 6.1 in. (Ankle High)
Height (Metric) 15.5 cm (Ankle High)
Resistance Liquid, Static
Product Type Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.