Learn More
Sigma Organic Chemistry MakroPor, silicon membrane | 10EA
Supplier: Sigma Organic Chemistry MP8475101010EA

MakroPor, silicon membrane, 10EA
About This Item:
Linear Formula: Si
UNSPSC Code: 12352200
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.