Learn More
Sigma Organic Chemistry MAL-dPEG(R)24-NHS ester | 100MG | MFCD11041160
Supplier: Sigma Organic Chemistry QBD10314100MG

MAL-dPEG(R)24-NHS ester, 100MG
About This Item:
Linear Formula: C62H111N3O31
MDL Number: MFCD11041160
UNSPSC Code: 12352200
Storage: -20C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.