Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1085 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1-085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1-085 Supplier Invitrogen™ Supplier No. PA1085
  • $596.37
    $461.65 / Each of 1

    Save $134.72

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-085 detects MAL from rat, mouse, xenopus and human samples. PA1-085 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~ 17 kDa protein corresponding to MAL from rat kidney extracts. The PA1-085 immunogen is a synthetic peptides corresponding to residues C(114)F D G F T Y (R/K) H Y H E N I(126) of human MAL. This peptide (Cat. # PEP-249) can be used in neutralization and control experiments.

MAL, a nonglycosylated integral membrane proteolipid, has been identified as a major internal component of glycolipid-enriched membrane (GEM) microdomains. MAL is the first integral membrane proteolipid component to have been shown as a necessary element in the cellular apical sorting apparatus for newly synthesized proteins. In its steady state this highly hydrophobic proteolipid, resides predominantly in the apical zone of polarized epithelial cells. It has also been shown in itinerant protein cycling between the plasma membrane and endosomes and in other cells. This 17-kDa proteolipid contains four putative transmembrane domains. These domains along with its protein trafficking properties are defined by the specific signals delegated by its specific amino acid sequence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAL
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene MAL
Gene Accession No. O09198, P21145, Q64349
Gene Alias 17 kDa myelin vesicular protein; Mal; mal T-cell differentiation protein; mal, T cell differentiation protein L homeolog; mal, T-cell differentiation protein; mal.L; MALGENE; Mpv17; Mvp17; MyD88-adapter-like; myelin and lymphocyte protein; myelin and lymphocyte protein, T cell differentiation protein; myelin and lymphocyte protein, T-cell differentiation protein; NS 3; T-lymphocyte maturation-associated protein; Vip17; XELAEV_18026637mg
Gene Symbols MAL, mal.L
Host Species Rabbit
Immunogen Synthetic peptides corresponding to residues C(114)F D G F T Y (R/K) H Y H E N I(126) of human MAL.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17153, 25263, 379738, 4118
Target Species Human, Mouse, Rat, Frog
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.