Learn More
Cayman Chemical Malachite Green Phosphate Assay Kit, 2 x 96 wells
SDP
Supplier:  Cayman Chemical 100093252X96WELLS
Encompass_Preferred
Caymans Malachite Green Phosphate Assay provides a fast, reproducible, colorimetric method for measuring inorganic free phosphate in aqueous solutions. The assay method is based on the formation of a complex between malachite green molybdate and free orthophosphate that absorbs at 620-640 nm. Applications for this assay include quantification of phosphorylation and phosphate release from protein phosphatase substrates. This assay is a reliable and suitable means of detecting and quantifying minimal amounts of inorganic free phosphate and is amenable to high-throughput screening applications. The assay is formatted to a 96-well format, but could easily be modified for use in 384-well or cuvette-based assays.
Catalog No. NC0028449
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.