Learn More
ATCC Malassezia globosa Midgley - Cryopreserved yeast culture, strain 2-2786 (ATCC MYA-4794)
Supplier:  ATCC MYA4794
Encompass
Malassezia globosa Midgley (ATCC MYA-4794) is a lipid-dependent yeast supplied as a cryopreserved viable culture for laboratory research. It is intended for reconstitution and propagation on lipid-enriched media for studies in fungal biology, dermatology, and microbiology. Handle and store according to biosafety guidelines.
  • An ampoule containing viable yeast cells suspended in cryoprotectant.
  • Recommended storage at -80°C or colder to maintain viability.
  • Grows on modified Dixon and Modified Leeming & Notman Agar media.
  • Incubate aerobically at 30-32°C for optimal growth.
  • Requires lipid supplementation for propagation.
Catalog No. NC3355792