Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD170412
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 6 4mm Polypropylene Overall Length 47 2 mm 1 86 in Almond Diameter Inner 0 25 in Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 0 89 in 22 6 mm
Catalog No. 50-313-1280
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.