Learn More
MilliporeSigma™ Supelco™ Male Luer Plug
Catalog No. 111020949 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
12/Pk.
Catalog No. Packaging Quantity
11-102-0949 12/Pk.
1 options
Catalog No. 11-102-0949 Supplier MilliporeSigma™ Supelco™ Supplier No. 504351
Add to Cart
Add to Cart
Male Luer plug is used to seal the inlet sampling cap which serves as an accessory for thermal desorption system storage containers.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 12/Pk.
Description For plugging luer holes