Learn More
Selleck Chemical LLC Indacaterol Maleate S3083-5mg
Supplier: Selleck Chemical LLC S30835MG
Indacaterol (QAB149) is an ultra-long-acting -adrenoceptor agonist with pKi of 7 36 for 1-adrenoceptor and pKi of 5 48 for 2-adrenoceptor
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.