Learn More
Nanocs MALEIMIDE PEG MPEG-MAL 10000DA
Supplier:  Nanocs PG1ML10K
Encompass_Preferred
Maleimide functionalized polyethylene glycol, Maleimide PEG is a sulfhydryl (thiol/SH) group reactive PEG derivative that can be used to selectively modify protein, peptide or any other surfaces with availablesulfhydryl groups. The reaction between maleimide and thiol proceeds readily at pH 6.5~7.5 to form stable thioether bonds.Store at -20 0C, dessiccated. Avoid frequent thaw and freeze.
Catalog No. NC2372807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.