Learn More
US Biomax Inc MALIGNANT MELANOMA TEST TISSUE
SDP
Supplier:  US Biomax Inc T382C/DEL
Encompass
Malignant melanoma test tissue array with skin tissue, including pathology grade, TNM and clinical stage, 12 cases/24 cores
Catalog No. NC2021782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.