Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MALP-2, TLR6 and TLR2 ligand
SDP

Catalog No. NBP226219 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226219 50 μg
NBP2262192 2μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226219 Supplier Novus Biologicals™ Supplier No. NBP226219

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Applications: Functional, In vitro assay

Specific Activity is approx. 2x10^8units/mg. One unit is defined as the dilution giving half maximal release of nitric oxide from C3H/HeJ mouse peritoneal exudate cells in the standard assay. Immunogen: MALP-2, TLR6 and TLR2 ligand Buffer: 2μg in 20μL of sterile PBS solution containing 2.5% (v/v) 2-propanol, 25mM n-octyl-b-D-glucopyranoside and 1% (wt/v) human serum albumin. Dilution: Functional, In vitro assay MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. MW: 2135.2

Specifications

Concentration 0.1 mg/mL
Formulation C99H167N19O30S
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Quantity 50 μg
For Use With (Application) Source: Synthetic Formula: C99H167N19O30S MW: 2135.2 Use in In-Vitro and Functional Assay Reported in Scientific Literature (PMID: 25466255).
Target MALP-2, TLR6 and TLR2 Ligand
Species Human/Mouse/Avian
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.