Learn More
Description
Specifications
Specifications
| Concentration | 0.1 mg/mL |
| Formulation | C99H167N19O30S |
| Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantity | 2μg |
| For Use With (Application) | Use in in vitro assay and functional reported in scientific literature (PMID 25466255) |
| Target | MALP-2, TLR6 and TLR2 Ligand |
| Species | Human/Mouse/Avian |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
