Learn More
Carbosynth LLC. Maltohexaose, 34620-77-4, MFCD00083650, 100 mg
SDP
Supplier:  Carbosynth LLC. OM06869100MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Chemical Formula: C36H62O31, MW: 990.86. 1,4-glucohexasaccharide derived from starch by hydrolysis and chromatography.
Catalog No. NC1106493
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.