Learn More
Medchemexpress LLC Maltopentaose | 34620-76-3 | 828.72 | C30H52O26 | 10 MG

Supplier: Medchemexpress LLC HYN149510MG
Maltopentaose is an analytical reference standard of maltopentaose (maltopentose) used as a substrate for amylases and as a reference for qualitative and quantitative analytical methods such as HPLC, GC, and MS. The material is supplied in small pack sizes with guidance for solubility and cold-storage handling.
- Analytical standard for HPLC, GC, and MS applications.
- Used as a substrate for amylase activity assays.
- Molecular formula C30H52O26; molecular weight 828.72.
- High aqueous solubility: water ≈116.67 mg/mL; PBS forms a clear solution at 100 mg/mL.
- Storage guidance: store at -20°C and protect from light; in solvent store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.