Learn More
Invitrogen™ Maltose Binding Protein Monoclonal Antibody (MBP61R)
Shop All Thermo Scientific Products

Description
Maltose Binding Protein Monoclonal Antibody for DB, ELISA, IP, Western Blot
Specifications
Specifications
| Antigen | Maltose Binding Protein |
| Applications | Dot Blot, ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | MBP61R |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide; pH 7.2 |
| Gene Alias | MBP; MBP Tag |
| Host Species | Mouse |
| Immunogen | Purified recombinant Maltose Binding fusion protein expressed in E.coli. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.