Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Maltose Binding Protein Recombinant Protein
SDP

Catalog No. NBC118538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118538 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118538 Supplier Novus Biologicals™ Supplier No. NBC118538
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 27-392 of E.coli Maltose Binding Protein The Maltose Binding Protein Recombinant Protein is derived from E. coli. The Maltose Binding Protein Recombinant Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.7 mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid in 20mM Tris pH 8.0
Gene ID (Entrez) 948538
Molecular Weight (g/mol) 42kDa
Name Maltose Binding Protein Protein
Purification Method Protein
Quantity 0.5 mg
Source E. Coli
Immunogen Maltose Binding Protein(MBP), 1-387 aa, E.coli, Recombinant, E.coli
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Bacteria
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.