Learn More
Pfanstiehl Inc Maltose Monohydrate, High Purity, Low Endotoxin, Low Metals, USP-NF, JP, ChP (M-167) 1kg, GMP , Multi-compendial CAS: 6363-53-7 Injectable Excipient Grade
Supplier:  Pfanstiehl Inc M1671KG
Encompass_Preferred
Pfanstiehl Maltose Monohydrate High Purity, Low Endotoxin, Low Metals is a multi-compendial carbohydrate utilized in bioprocessing applications and as an infusion stabilizer. Pfanstiehl Maltose has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Maltose meets parenteral (injectable) excipient grade standards, making it ideal as a bioprocessing feed, or as a stabilizing agent for pharmaceutical infusions. Pfanstiehl Maltose is available as USP-NF (National Formulary) JP (Japanese Pharmacopeia) and ChP (Chinese Pharmacopeia) Compendial, U.S.P. , cGMP , pharmaceutical grade .
Catalog No. NC2063107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.