Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltotriose, 93%

Catalog No. AC225940010
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225940010 1 g
AC225945000 500 mg
AC225940250 25 g
AC225940050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC225940010 Supplier Thermo Scientific Chemicals Supplier No. 225940010

CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1109-28-0
MDL Number MFCD00006629
InChI Key FYGDTMLNYKFZSV-NBCPLHMPSA-N
Synonym maltotriose, d-maltotriose
PubChem CID 134129496
IUPAC Name (2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 93%
Molecular Formula C18H32O16
Quantity 1 g
Specific Rotation Condition +162.50° (25°C c=1,H2O)
Specific Rotation +160° to +165° (25°C, 589nm) (c=1, H2O)
Molecular Weight (g/mol) 504.438
Formula Weight 504.44
Percent Purity ≥91%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material Maltotriose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.