Learn More
Eisco™ Malvern Eddy Current Unit
SCI_ED
Catalog No. S15123 Shop All Eisco Products
Change view
Click to view available options
Malvern Eddy Current Unit
Catalog No.
S15123 Malvern Eddy Current Unit
1 options
Catalog No. S15123 Supplier Eisco™ Supplier No. PH0467J
Add to Cart
Add to Cart
Malvern Eddy Current Unit for the demonstration of eddy current braking when used in conjunction with a hand wheel driving unit.
  • For the demonstration of eddy current braking when used in conjunction with the hand wheel driving unit
  • Powerful circular magnets are mounted on a pivoted arm so that they may be moved over or away from the disc as required
  • Base has rubber stoppers underneath to prevent the apparatus from moving or sliding on the surface while in use
  • Mounted on a secure wooden base
  • Aluminum disc has a 4.5 in. diameter, and a 20 mm driving pulley
  • Overall apparatus measures 7 x 4.6 x 6.5 in.
Specifications
Product Type Malvern Eddy Current Unit
Description Energy Transfer Apparatus
Material Aluminum (Disc), Wood (Base)
For Use With (Application) Demonstration of Eddy Current Braking
Height (English) 6.5 in.