Learn More
Eisco™ Malvern Motor/Generator
SCI_ED
Catalog No. S15120 Shop All Eisco Products
Change view
Click to view available options
Malvern Motor
Catalog No.
S15120 Malvern Motor
1 options
Catalog No. S15120 Supplier Eisco™ Supplier No. PH0467A
Add to Cart
Add to Cart
The Malvern Motor enables users to qualitatively show energy conversion from one form to another in a variety of ways.
  • Ideal for use as a tool when studying the input/output of larger motors on a comparative level
  • Use in conjunction with the Malvern Small Motor/Generator (PH0467B)
  • Mounted driving unit or dynamo
  • 2 to 6V DC motor and 15 mm V pulley
  • Mounted on base with 4 mm terminal socket
  • Base measures 6 x 4 in.
Specifications
Product Type Malvern Motor
Description Energy Transfer Apparatus
For Use With (Application) Qualitative Depiction of Energy Conversion
Includes 15 mm V-Pulley, 2 to 6 V D.C. Motor