Learn More
MAML2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300681AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.Specifications
| MAML2 | |
| Polyclonal | |
| Unconjugated | |
| MAML2 | |
| Mastermind-like protein 2, Mam-2, MAML2, KIAA1819 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84441 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8IZL2 | |
| MAML2 | |
| Between 250 and 300 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.